Learn More
Medchemexpress LLC HY-134818 5mg , ML-SI1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1348185MG
Encompass_Preferred
HY-134818 5mg ML-SI1 CAS: ML-SI1, a racemic mixture of diastereomers, is a TRPML inhibitor with an IC50 value of 15 μM for TRPML1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8991