Learn More
Medchemexpress LLC HY-100230A 5mg Medchemexpress, ML133 (hydrochloride) CAS:1222781-70-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100230A5MG
Encompass_Preferred
Medchemexpress, HY-100230A 5mg ML133 (hydrochloride) CAS:1222781-70-5 ML133 hydrochloride is a selective K ir 2 family channels inhibitor, with an IC 50 of 1.8 μM at pH 7.4 and 290 nM at pH 8.5 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1766