Learn More
Medchemexpress LLC HY-12755 10mg , ML141 CAS:71203-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1275510MG
Encompass_Preferred
HY-12755 10mg ML141 CAS: 71203-35-5 ML141 (CID-2950007) is a potent, allosteric, selective and reversible non-competitive inhibitor of Cdc42 GTPase. ML141 inhibits Cdc42 wild type and Cdc42 Q61L mutant with EC50s of 2.1 and 2.6 μM, respectively. ML141 shows low micromolar potency and selectivity against other members of the Rho family of GTPases (Rac1, Rab2, Rab7). ML141 do not show cytotoxicity in multiple cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9665