Learn More
Selleck Chemical LLC ML141 5mg 71203-35-5 CID-2950007
SDP
Supplier:  Selleck Chemical LLC S76865MG
Encompass_Preferred
ML141 (CID-2950007) is demonstrated to be a potent, selective and reversible non-competitive inhibitor of Cdc42 GTPase suitable for in vitro assays, with IC50 of 200 nM and selectivity against other members of the Rho family of GTPases (Rac1, Rab2, Rab7). ML141 is associated with an increase in p38 activation and may induce p38-dependent apoptosis/senescence. ML141 also protects neuroblastoma cells from metformin-induced apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5131