Learn More
Selleck Chemical LLC ML161 (Parmodulin 2) S2916-10mg
SDP
Catalog No. 503111694
Encompass_Preferred
ML161 (Parmodulin 2) is an allosteric inhibitor of PAR1 with IC50 of 0 26 M
Catalog No. 50-311-1694 Supplier Selleck Chemical LLC Supplier No. S291610MG
ML161 (Parmodulin 2) is an allosteric inhibitor of PAR1 with IC50 of 0 26 M