Learn More
TARGETMOL CHEMICALS INC ML162 10MG
SDP
Catalog No. 502579033
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML162 is an inhibitor of GPX4 that is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50s = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts respectively)purity: 99%
Catalog No. 50-257-9033 Supplier TARGETMOL CHEMICALS INC Supplier No. T897010MG
Also available in 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML162 is an inhibitor of GPX4 that is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50s = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts respectively)purity: 99%