Learn More
Selleck Chemical LLC ML162 5mg
SDP
Supplier:  Selleck Chemical LLC S44525MG
Encompass_Preferred
ML162 is a covalent inhibitor of cellular phospholipid glutathione peroxidase (GPX-4) that induces ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3143