Learn More
Medchemexpress LLC ML162-yne | 2883115-46-4 | 99.1% | 501.42 g·mol⁻¹ | C25H22Cl2N2O3S | 10 MG
SDP
Catalog No. 5000286094
Encompass_Preferred
ML162-yne is a GPX4-inhibitor affinity probe designed for click-chemistry applications. The molecule contains an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for labeling or capturing target-engaged proteins. It is supplied as a high-purity solid or as a DMSO solution and is intended for research use only.
  • Enables selective labeling via copper-catalyzed azide-alkyne cycloaddition.
  • Functions as an affinity probe for glutathione peroxidase 4 (GPX4).
  • Supplied with high purity for reliable experimental results.
  • Available as solid and ready-to-use DMSO solution formats.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-86094 Supplier Medchemexpress LLC Supplier No. HY15374810MG
ML162-yne is a GPX4-inhibitor affinity probe designed for click-chemistry applications. The molecule contains an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for labeling or capturing target-engaged proteins. It is supplied as a high-purity solid or as a DMSO solution and is intended for research use only.
  • Enables selective labeling via copper-catalyzed azide-alkyne cycloaddition.
  • Functions as an affinity probe for glutathione peroxidase 4 (GPX4).
  • Supplied with high purity for reliable experimental results.
  • Available as solid and ready-to-use DMSO solution formats.
  • Stable under recommended storage conditions for long-term use.