Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ML162-yne | 2883115-46-4 | C₂₅H₂₂Cl₂N₂O₃S | 25 MG
SDP

Catalog No. 5000375184
Encompass_Preferred

ML162-yne is a potent GPX4-inhibitor affinity probe. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.

  • Potent GPX4-inhibitor affinity probe.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Purity: 99.06%.

Catalog No. 50-003-75184 Supplier Medchemexpress LLC Supplier No. 25MGHY153748
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML162-yne is a potent GPX4-inhibitor affinity probe. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.

  • Potent GPX4-inhibitor affinity probe.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Purity: 99.06%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.