Learn More
Medchemexpress LLC ML162-yne | 2883115-46-4 | C25H22Cl2N2O3S | 10 MM 1 ML
SDP
Catalog No. 5000376246
Encompass_Preferred
ML162-yne is a potent GPX4-inhibitor affinity probe. It is a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent GPX4 inhibitor
  • Affinity probe
  • Click chemistry reagent
  • Contains alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-76246 Supplier Medchemexpress LLC Supplier No. HY15374810MM/1ML
ML162-yne is a potent GPX4-inhibitor affinity probe. It is a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent GPX4 inhibitor
  • Affinity probe
  • Click chemistry reagent
  • Contains alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)