Learn More
Medchemexpress LLC ML162-yne | 2883115-46-4 | C25H22Cl2N2O3S | 100 MG
SDP
Catalog No. 5000376024
Encompass_Preferred
ML162-yne is a potent GPX4-inhibitor affinity probe. It is a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent GPX4-inhibitor affinity probe
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-76024 Supplier Medchemexpress LLC Supplier No. HY153748100MG
ML162-yne is a potent GPX4-inhibitor affinity probe. It is a click chemistry reagent that contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent GPX4-inhibitor affinity probe
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups