Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ML162-yne | 2883115-46-4 | C25H22Cl2N2O3S | 5 MG
SDP

Catalog No. 5000376770
Encompass_Preferred

ML162-yne is a potent GPX4-inhibitor affinity probe and a click chemistry reagent. It contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.

  • Potent GPX4-inhibitor affinity probe
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • High purity: 99.06%
  • Molecular weight: 501.42
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL)
  • Targets GPX4

Catalog No. 50-003-76770 Supplier Medchemexpress LLC Supplier No. HY1537485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML162-yne is a potent GPX4-inhibitor affinity probe and a click chemistry reagent. It contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.

  • Potent GPX4-inhibitor affinity probe
  • Click chemistry reagent
  • Contains an Alkyne group for CuAAc reactions
  • High purity: 99.06%
  • Molecular weight: 501.42
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL)
  • Targets GPX4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.