Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ML162-yne | 2883115-46-4 | C25H22Cl2N2O3S | 50 MG
SDP

Catalog No. 5000376300
Encompass_Preferred

ML162-yne is a potent GPX4-inhibitor affinity probe and a click chemistry reagent. It contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.

  • Potent GPX4 inhibitor
  • Affinity probe
  • Click chemistry reagent
  • Contains alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Purity of 99.06% (LCMS)
  • Appearance: white to off-white solid
  • Used in cancer research
  • Targeted therapy for cancer
  • Relevant to cancer metabolism and metastasis studies
  • Involved in apoptosis research
  • Acts as a metabolic enzyme/protease inhibitor
  • Inhibits glutathione peroxidase

Catalog No. 50-003-76300 Supplier Medchemexpress LLC Supplier No. HY15374850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML162-yne is a potent GPX4-inhibitor affinity probe and a click chemistry reagent. It contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.

  • Potent GPX4 inhibitor
  • Affinity probe
  • Click chemistry reagent
  • Contains alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Purity of 99.06% (LCMS)
  • Appearance: white to off-white solid
  • Used in cancer research
  • Targeted therapy for cancer
  • Relevant to cancer metabolism and metastasis studies
  • Involved in apoptosis research
  • Acts as a metabolic enzyme/protease inhibitor
  • Inhibits glutathione peroxidase

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.