Learn More
Apexbio Technology LLC ML162-YNE-5MG
SDP
Catalog No. 5000570029
Encompass_Preferred
ML162-yne (CAS No 2883115-46-4) is a compound predominantly used in biomedical research as a kinase inhibitor Originating in specialized laboratories focused on signal transduction pathways ML162-yne acts by targeting specific kinases involved in cellular signaling thereby modulating critical biochemical pathways Its in vitro activity is observed in the nanomolar to low micromolar range making it a potent agent for studies investigating kinase-mediated processes Researchers commonly utilize ML162-yne in both cellular and animal models to elucidate its effect on cell proliferation apoptosis and other vital cellular functions This compound is frequently used in drug screening assays to discover and optimize new therapeutic agents targeting kinase pathways Experimental concentrations of ML162-yne typically depend on the specific research design allowing flexibility in its application across various investigational contexts
Catalog No. 50-005-70029 Supplier Apexbio Technology LLC Supplier No. BA34485
ML162-yne (CAS No 2883115-46-4) is a compound predominantly used in biomedical research as a kinase inhibitor Originating in specialized laboratories focused on signal transduction pathways ML162-yne acts by targeting specific kinases involved in cellular signaling thereby modulating critical biochemical pathways Its in vitro activity is observed in the nanomolar to low micromolar range making it a potent agent for studies investigating kinase-mediated processes Researchers commonly utilize ML162-yne in both cellular and animal models to elucidate its effect on cell proliferation apoptosis and other vital cellular functions This compound is frequently used in drug screening assays to discover and optimize new therapeutic agents targeting kinase pathways Experimental concentrations of ML162-yne typically depend on the specific research design allowing flexibility in its application across various investigational contexts