Learn More
Medchemexpress LLC HY-15951 5mg Medchemexpress, ML167 CAS:1285702-20-6 Purity:>98%
SDP
Catalog No. 502081846
Encompass_Preferred
Medchemexpress, HY-15951 5mg ML167 CAS:1285702-20-6 ML167 is a highly selective Cdc2-like kinase 4 (Clk4) inhibitor with IC50 of 136 nM, >10-fold selectivity for closely related kinases Clk1, Clk2, Clk3 and Dyrk1A/1B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1846 Supplier Medchemexpress LLC Supplier No. HY159515MG
Medchemexpress, HY-15951 5mg ML167 CAS:1285702-20-6 ML167 is a highly selective Cdc2-like kinase 4 (Clk4) inhibitor with IC50 of 136 nM, >10-fold selectivity for closely related kinases Clk1, Clk2, Clk3 and Dyrk1A/1B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.