Learn More
Medchemexpress LLC HY-12805 100mg Medchemexpress, ML171 CAS:6631-94-3 Purity:>98%
SDP
Catalog No. 502081148
Encompass_Preferred
Medchemexpress, HY-12805 100mg ML171 CAS:6631-94-3 ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1148 Supplier Medchemexpress LLC Supplier No. HY12805100MG
Medchemexpress, HY-12805 100mg ML171 CAS:6631-94-3 ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.