Learn More
Medchemexpress LLC ML204 hydrochloride | 2070015-10-8 | 99.7% | 1 ML
SDP
Catalog No. 5000340168
Encompass_Preferred
ML204 hydrochloride is a novel, potent, and selective TRPC4/TRPC5 channel inhibitor.
  • Potent and selective TRPC4/TRPC5 channel inhibitor.
  • Exhibits at least 19-fold selectivity against TRPC6.
  • No appreciable effect on other TRP channels.
  • Does not affect voltage-gated sodium, potassium, or Ca2+ channels.
Catalog No. 50-003-40168 Supplier Medchemexpress LLC Supplier No. HY12949A10MM/1ML
ML204 hydrochloride is a novel, potent, and selective TRPC4/TRPC5 channel inhibitor.
  • Potent and selective TRPC4/TRPC5 channel inhibitor.
  • Exhibits at least 19-fold selectivity against TRPC6.
  • No appreciable effect on other TRP channels.
  • Does not affect voltage-gated sodium, potassium, or Ca2+ channels.