Learn More
Medchemexpress LLC HY-12949A 5mg Medchemexpress, ML204 (hydrochloride) CAS:2070015-10-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12949A5MG
Encompass_Preferred
Medchemexpress, HY-12949A 5mg ML204 (hydrochloride) CAS:2070015-10-8 ML204 hydrochloride is a novel, potent, selective TRPC4/TRPC5 channel inhibitor, with at least 19-fold selectivity against TRPC6 and no appreciable effect on all other TRP channels, nor on voltage-gated sodium, potassium, or Ca2+ channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1211