Learn More
Medchemexpress LLC HY-101843 5mg Medchemexpress, ML213 CAS:489402-47-3 Purity:>98%
SDP
Catalog No. 501967884
Encompass_Preferred
Medchemexpress, HY-101843 5mg ML213 CAS:489402-47-3 ML213 is a selective activator of Kv7.2 and Kv7.4 channels, enhances Kv7.2 and Kv7.4 channels with EC50s of 230 and 510 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7884 Supplier Medchemexpress LLC Supplier No. HY1018435MG
Medchemexpress, HY-101843 5mg ML213 CAS:489402-47-3 ML213 is a selective activator of Kv7.2 and Kv7.4 channels, enhances Kv7.2 and Kv7.4 channels with EC50s of 230 and 510 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.