Learn More
Apexbio Technology LLC ML216 1430213-30-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063630
Encompass_Preferred
ML216 (CAS 1430213-30-1) is a small molecule inhibitor targeting Bloom (BLM) helicase a DNA unwinding enzyme essential for homologous recombination-mediated DNA repair ML216 inhibits DNA unwinding activity of full-length BLM (IC50 3 0 M) and a truncated form BLM636-1298 (IC50 0 97 M) displaying sub-micromolar selectivity relative to other helicases such as RECQ1 RECQL5 and bacterial UvrD In cellular assays ML216 selectively suppresses proliferation in BLM-containing fibroblasts increasing sister chromatid exchange frequency consistent with BLM functional deficiency By inhibiting BLM helicase ML216 may sensitize tumor cells to conventional DNA-damaging therapeutics indicating potential application in cancer research and therapy development
Catalog No. 50-000-63630 Supplier Apexbio Technology LLC Supplier No. B80155.1
ML216 (CAS 1430213-30-1) is a small molecule inhibitor targeting Bloom (BLM) helicase a DNA unwinding enzyme essential for homologous recombination-mediated DNA repair ML216 inhibits DNA unwinding activity of full-length BLM (IC50 3 0 M) and a truncated form BLM636-1298 (IC50 0 97 M) displaying sub-micromolar selectivity relative to other helicases such as RECQ1 RECQL5 and bacterial UvrD In cellular assays ML216 selectively suppresses proliferation in BLM-containing fibroblasts increasing sister chromatid exchange frequency consistent with BLM functional deficiency By inhibiting BLM helicase ML216 may sensitize tumor cells to conventional DNA-damaging therapeutics indicating potential application in cancer research and therapy development