Learn More
Selleck Chemical LLC ML218 E8077-1MG
SDP
Catalog No. 5000042411
Encompass_Preferred
ML218 is a potent selective and orally active T-type Ca2 channels (Cav3 1 Cav3 2 Cav3 3) inhibitor with IC50s of 310 nM and 270 nM for Cav3 2 and Cav3 3 respectively ML218 inhibits the burst activity in subthalamic nucleus (STN) neurons ML218 has no significant inhibition of L- or N-type calcium channels KATP or hERG potassium channels ML218 can penetrate the blood-brain barrier
Catalog No. 50-000-42411 Supplier Selleck Chemical LLC Supplier No. E80771MG
ML218 is a potent selective and orally active T-type Ca2 channels (Cav3 1 Cav3 2 Cav3 3) inhibitor with IC50s of 310 nM and 270 nM for Cav3 2 and Cav3 3 respectively ML218 inhibits the burst activity in subthalamic nucleus (STN) neurons ML218 has no significant inhibition of L- or N-type calcium channels KATP or hERG potassium channels ML218 can penetrate the blood-brain barrier