Learn More
TARGETMOL CHEMICALS INC ML221 25MG
SDP
Catalog No. 502579888
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML221 is a potent apelin /APJ functional antagonist inhibiting apelin-13-mediated activation of APJ with IC50s of 0.70 uM in the cAMP assay and 1.75 uM in the (beta)-arrestin assay and EC80 of 10 nM in both assays. purity: 97%
Catalog No. 50-257-9888 Supplier TARGETMOL CHEMICALS INC Supplier No. T439025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML221 is a potent apelin /APJ functional antagonist inhibiting apelin-13-mediated activation of APJ with IC50s of 0.70 uM in the cAMP assay and 1.75 uM in the (beta)-arrestin assay and EC80 of 10 nM in both assays. purity: 97%