Learn More
Medchemexpress LLC HY-103254 5mg Medchemexpress, ML221 CAS:877636-42-5 Purity:>98%
SDP
Catalog No. 501968062
Encompass_Preferred
Medchemexpress, HY-103254 5mg ML221 CAS:877636-42-5 ML221 is a potent apelin (APJ) functional antagonist, inhibiting apelin-13-mediated activation of APJ, with IC50s of 0.70 μM in the cAMP assay, and 1.75 μM in the β-arrestin assay, and EC80 of 10 nM in both assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8062 Supplier Medchemexpress LLC Supplier No. HY1032545MG
Medchemexpress, HY-103254 5mg ML221 CAS:877636-42-5 ML221 is a potent apelin (APJ) functional antagonist, inhibiting apelin-13-mediated activation of APJ, with IC50s of 0.70 μM in the cAMP assay, and 1.75 μM in the β-arrestin assay, and EC80 of 10 nM in both assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.