Learn More
Medchemexpress LLC Ml224 (NCGC00242364) | 1338824-21-7 | 99.2% | 525.59 g·mol⁻¹ | C31H31N3O5 | 50MG
SDP
Catalog No. 5000239671
Encompass_Preferred
ML224 (NCGC00242364) is a small-molecule selective antagonist of the thyroid-stimulating hormone receptor (TSHR) with an IC50 of approximately 2.1 μM. It is supplied as a high-purity research reagent for in vitro pharmacology and biochemical studies and is used to investigate Graves' disease and other thyroid disorders.
  • Selective TSHR antagonist with IC50 ≈ 2.1 μM.
  • High purity suitable for in vitro studies.
  • Molecular formula C31H31N3O5 and molecular weight 525.59 g·mol⁻¹.
  • Available as solid powder and as DMSO solution formats.
  • Intended for pharmacology and cell-based assay applications.
This item is not returnable. View return policy
Catalog No. 50-002-39671 Supplier Medchemexpress LLC Supplier No. HY1238150MG
ML224 (NCGC00242364) is a small-molecule selective antagonist of the thyroid-stimulating hormone receptor (TSHR) with an IC50 of approximately 2.1 μM. It is supplied as a high-purity research reagent for in vitro pharmacology and biochemical studies and is used to investigate Graves' disease and other thyroid disorders.
  • Selective TSHR antagonist with IC50 ≈ 2.1 μM.
  • High purity suitable for in vitro studies.
  • Molecular formula C31H31N3O5 and molecular weight 525.59 g·mol⁻¹.
  • Available as solid powder and as DMSO solution formats.
  • Intended for pharmacology and cell-based assay applications.