Learn More
Medchemexpress LLC HY-12754 5mg Medchemexpress, ML228 CAS:1357171-62-0 Purity:>98%
SDP
Catalog No. 502081135
Encompass_Preferred
Medchemexpress, HY-12754 5mg ML228 CAS:1357171-62-0 ML228 (CID-46742353) is a potent the Hypoxia Inducible Factor (HIF) pathway activator with EC50 of 1 μM. ML228 potently activates HIF in vitro as well as its downstream target VEGF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1135 Supplier Medchemexpress LLC Supplier No. HY127545MG
Medchemexpress, HY-12754 5mg ML228 CAS:1357171-62-0 ML228 (CID-46742353) is a potent the Hypoxia Inducible Factor (HIF) pathway activator with EC50 of 1 μM. ML228 potently activates HIF in vitro as well as its downstream target VEGF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.