Learn More
Medchemexpress LLC HY-19971 5mg Medchemexpress, ML239 CAS:1378872-36-6 Purity:>98%
SDP
Catalog No. 502082204
Encompass_Preferred
Medchemexpress, HY-19971 5mg ML239 CAS:1378872-36-6 ML239 is a potent and selective inhibitor of breast cancer stem cells, with an IC50 of 1.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2204 Supplier Medchemexpress LLC Supplier No. HY199715MG
Medchemexpress, HY-19971 5mg ML239 CAS:1378872-36-6 ML239 is a potent and selective inhibitor of breast cancer stem cells, with an IC50 of 1.16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.