Learn More
Medchemexpress LLC HY-19797A 100mg , ML241 (hydrochloride) CAS: Purity:98%
SDP
Catalog No. 501493192
Encompass_Preferred
Medchemexpress, HY-19797A 100mg ML241 (hydrochloride) CAS: Formula:C23H25ClN4O IC50: 100 nM (p97) Purity:98% ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC 50 value of 100 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3192 Supplier Medchemexpress LLC Supplier No. HY19797A100MG
Add to Cart
Add to Cart
Medchemexpress, HY-19797A 100mg ML241 (hydrochloride) CAS: Formula:C23H25ClN4O IC50: 100 nM (p97) Purity:98% ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC 50 value of 100 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.