Learn More
Abcam ML297 (VU0456810), GIRK activator, 100MG
Catalog No. 502706108
Encompass_Preferred
MW 328.32 Da, Purity >99%. Potent, selective GIRK activator (EC₅₀ = 160 nM). Shows anticonvulsant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6108 Supplier Abcam Supplier No. AB143564100MG
MW 328.32 Da, Purity >99%. Potent, selective GIRK activator (EC₅₀ = 160 nM). Shows anticonvulsant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement