Learn More
Medchemexpress LLC HY-101778 100mg Medchemexpress, ML311 CAS:315698-17-0 Purity:>98%
SDP
Catalog No. 501932211
Encompass_Preferred
Medchemexpress, HY-101778 100mg ML311 CAS:315698-17-0 ML311 is a potent and selective inhibitor of the Mcl-1/Bim interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2211 Supplier Medchemexpress LLC Supplier No. HY101778100MG
Medchemexpress, HY-101778 100mg ML311 CAS:315698-17-0 ML311 is a potent and selective inhibitor of the Mcl-1/Bim interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.