Learn More
Selleck Chemical LLC ML323 5mg
SDP
Supplier:  Selleck Chemical LLC S75295MG
Encompass_Preferred
ML323 displays reversible, nanomolar inhibitory activity and excellent selectivity toward USP1/UAF1 with IC50 of 76 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3283