Learn More
Apexbio Technology LLC ML324 1222800-79-4 200mg
SDP
Catalog No. 5000092774
Encompass_Preferred
ML324 (CAS 1222800-79-4) is a small-molecule inhibitor targeting JMJD2 histone demethylase enzymes It is designed to suppress JMJD2-mediated demethylation of histone H3 lysine 9 (H3K9) thereby affecting gene transcription regulation ML324 exerts its biological activity primarily through inhibition of JMJD2 demethylases In cell-based studies ML324 inhibits JMJD2 activity displaying an IC50 value of 920 nM In experimental models including HFF and MRC-5 cell lines it blocks immediate-early (IE) gene expression of HSV and hCMV reduces viral spread and plaque formation and inhibits HSV-1 reactivation and replication in latent infection models such as mouse neuronal ganglia explants Based on these pharmacological properties ML324 holds research potential in chromatin remodeling and antiviral therapeutic strategies
Catalog No. 50-000-92774 Supplier Apexbio Technology LLC Supplier No. B4891200
ML324 (CAS 1222800-79-4) is a small-molecule inhibitor targeting JMJD2 histone demethylase enzymes It is designed to suppress JMJD2-mediated demethylation of histone H3 lysine 9 (H3K9) thereby affecting gene transcription regulation ML324 exerts its biological activity primarily through inhibition of JMJD2 demethylases In cell-based studies ML324 inhibits JMJD2 activity displaying an IC50 value of 920 nM In experimental models including HFF and MRC-5 cell lines it blocks immediate-early (IE) gene expression of HSV and hCMV reduces viral spread and plaque formation and inhibits HSV-1 reactivation and replication in latent infection models such as mouse neuronal ganglia explants Based on these pharmacological properties ML324 holds research potential in chromatin remodeling and antiviral therapeutic strategies