Learn More
Medchemexpress LLC HY-101464 5mg Medchemexpress, ML329 CAS:19992-50-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014645MG
Encompass_Preferred
Medchemexpress, HY-101464 5mg ML329 CAS:19992-50-8 ML329 is a small molecule inhibitor of MITF ; inhibits TRPM-1 promoter activity with an IC 50 of 1.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2141