Learn More
Medchemexpress LLC ML334 | 1432500-66-7 | 98.1% | 446.50 | 1 ML
SDP
Catalog No. 5000344228
Encompass_Preferred
ML334 is a potent, cell-permeable activator of NRF2, functioning by inhibiting the Keap1-NRF2 protein-protein interaction. It binds to the Keap1 Kelch domain with a Kd of 1 μM. ML334 stimulates NRF2 expression and nuclear translocation, leading to the induction of antioxidant response elements (ARE) activity.
  • Potent, cell permeable activator of NRF2.
  • Inhibits Keap1-NRF2 protein-protein interaction.
  • Binds to Keap1 Kelch domain with a Kd of 1 μM.
  • Stimulates NRF2 expression and nuclear translocation.
  • Induces antioxidant response elements (ARE) activity.
  • Increases NQO1 and TRX1 mRNAs in HEK293 cells.
  • Enhances HO-1 mRNA expression in HEK293 cells.
  • Induces HO-1 and TRX1 proteins in HEK293 cells.
Catalog No. 50-003-44228 Supplier Medchemexpress LLC Supplier No. HY11025810MG
ML334 is a potent, cell-permeable activator of NRF2, functioning by inhibiting the Keap1-NRF2 protein-protein interaction. It binds to the Keap1 Kelch domain with a Kd of 1 μM. ML334 stimulates NRF2 expression and nuclear translocation, leading to the induction of antioxidant response elements (ARE) activity.
  • Potent, cell permeable activator of NRF2.
  • Inhibits Keap1-NRF2 protein-protein interaction.
  • Binds to Keap1 Kelch domain with a Kd of 1 μM.
  • Stimulates NRF2 expression and nuclear translocation.
  • Induces antioxidant response elements (ARE) activity.
  • Increases NQO1 and TRX1 mRNAs in HEK293 cells.
  • Enhances HO-1 mRNA expression in HEK293 cells.
  • Induces HO-1 and TRX1 proteins in HEK293 cells.