Learn More
Medchemexpress LLC HY-110258 5mg Medchemexpress, ML334 CAS:1432500-66-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102585MG
Encompass_Preferred
Medchemexpress, HY-110258 5mg ML334 CAS:1432500-66-7 ML334 is a potent, cell permeable activator of NRF2 by inhibition of Keap1-NRF2 protein-protein interaction. ML334 binds to Keap1 Kelch domain with a Kd of 1 μM. ML334 stimulates NRF2 expression and nuclear translocation and induces antioxidant response elements (ARE) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8781