Learn More
Medchemexpress LLC HY-104005 100mg Medchemexpress, ML335 CAS:825658-06-8 Purity:98%
SDP
Catalog No. 501637223
Encompass_Preferred
Medchemexpress,HY-104005 100mg ML335 CAS:825658-06-8 Formula:C15H14Cl2N2O3S ML335 is a selective activator of both TREK-1 and TREK-2. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7223 Supplier Medchemexpress LLC Supplier No. HY104005100MG
Medchemexpress,HY-104005 100mg ML335 CAS:825658-06-8 Formula:C15H14Cl2N2O3S ML335 is a selective activator of both TREK-1 and TREK-2. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.