Learn More
Apexbio Technology LLC ML347 1062368-49-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062838
Encompass_Preferred
ML347 (CAS 1062368-49-3) is a potent and selective inhibitor of bone morphogenetic protein (BMP) type I receptors specifically targeting ALK2 Mechanistically ML347 selectively inhibits ALK2 kinase activity with an IC50 of 32 nM while also inhibiting ALK1 with a comparable IC50 of 46 nM Notably ML347 displays approximately 300-fold selectivity for ALK2 over ALK3 and lacks inhibitory activity against closely related kinases such as ALK6 and KDR In the BMP4-induced C2C12BRA cell-based assay ML347 exhibits inhibitory activity with an IC50 of 152 nM As a selective ALK2 inhibitor ML347 serves as a valuable probe in BMP pathway research
Catalog No. 50-000-62838 Supplier Apexbio Technology LLC Supplier No. B36885.1
ML347 (CAS 1062368-49-3) is a potent and selective inhibitor of bone morphogenetic protein (BMP) type I receptors specifically targeting ALK2 Mechanistically ML347 selectively inhibits ALK2 kinase activity with an IC50 of 32 nM while also inhibiting ALK1 with a comparable IC50 of 46 nM Notably ML347 displays approximately 300-fold selectivity for ALK2 over ALK3 and lacks inhibitory activity against closely related kinases such as ALK6 and KDR In the BMP4-induced C2C12BRA cell-based assay ML347 exhibits inhibitory activity with an IC50 of 152 nM As a selective ALK2 inhibitor ML347 serves as a valuable probe in BMP pathway research