Learn More
Selleck Chemical LLC ML347 5mg 1062368-49-3 LDN-193719
SDP
Supplier:  Selleck Chemical LLC S71485MG
Encompass_Preferred
ML347 (LDN-193719) is a selective BMP receptor inhibitor with IC50 of 32 nM for ALK2, >300-fold selectivity over ALK3. Also inhibits ALK1 activity with IC50 of 46 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8131