Learn More
Medchemexpress LLC HY-100736 5mg Medchemexpress, ML348 CAS:899713-86-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007365MG
Encompass_Preferred
Medchemexpress, HY-100736 5mg ML348 CAS:899713-86-1 ML348 is a selective and reversible lysophospholipase 1 (LYPLA1) inhibitor with an IC 50 of 210 nM, and barely inhibits LYPLA2 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1932