Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(1,3-benzothiazol-2-yl)-4-[(2-hydroxy-3-methoxyphenyl)methylamino]benzenesulfonamide | 1532593-30-8 | 98.1% | 441.52 g/mol | C21H19N3O4S2 | 5MG
SDP

Catalog No. 502081016
Encompass_Preferred

ML355 is a selective small-molecule inhibitor of 12-lipoxygenase (12-LOX) used for biochemical and cellular research. It inhibits 12-LOX with an IC50 of 0.34 μM, shows selectivity over related lipoxygenases and cyclooxygenases, and is suitable for in vitro assays and in vivo studies when prepared in recommended formulations.

  • Selective 12-LOX inhibition (IC50 0.34 μM).
  • High reported purity (98.1%).
  • Molecular weight 441.52 g/mol.
  • Soluble in DMSO (≥ 42 mg/mL); compatible with published in vivo formulations.
  • Solid form with recommended storage at -20 °C for long-term stability.

Catalog No. 50-208-1016 Supplier Medchemexpress LLC Supplier No. HY123415MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML355 is a selective small-molecule inhibitor of 12-lipoxygenase (12-LOX) used for biochemical and cellular research. It inhibits 12-LOX with an IC50 of 0.34 μM, shows selectivity over related lipoxygenases and cyclooxygenases, and is suitable for in vitro assays and in vivo studies when prepared in recommended formulations.

  • Selective 12-LOX inhibition (IC50 0.34 μM).
  • High reported purity (98.1%).
  • Molecular weight 441.52 g/mol.
  • Soluble in DMSO (≥ 42 mg/mL); compatible with published in vivo formulations.
  • Solid form with recommended storage at -20 °C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.