Learn More
TARGETMOL CHEMICALS INC ML364 25MG
SDP
Catalog No. 502580215
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML364 is an inhibitor of ubiquitin specific peptidase 2 (USP2) and can be used for the research of breast cancer. purity: 99%
Catalog No. 50-258-0215 Supplier TARGETMOL CHEMICALS INC Supplier No. T355525MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML364 is an inhibitor of ubiquitin specific peptidase 2 (USP2) and can be used for the research of breast cancer. purity: 99%