Learn More
Medchemexpress LLC HY-100523 10mg Medchemexpress, ML385 CAS:846557-71-9 Purity:>98%
SDP
Catalog No. 502028626
Encompass_Preferred
Medchemexpress, HY-100523 10mg ML385 CAS:846557-71-9 ML385 is a specific nuclear factor erythroid 2-related factor 2 (NRF2) inhibitor with an IC50 of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8626 Supplier Medchemexpress LLC Supplier No. HY10052310MG
Medchemexpress, HY-100523 10mg ML385 CAS:846557-71-9 ML385 is a specific nuclear factor erythroid 2-related factor 2 (NRF2) inhibitor with an IC50 of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.