Learn More
Apexbio Technology LLC ML385(Synonyms: ML-385, NRF2 inhibitor ML385, ML385 NRF2 inhibitor, ML385 inhibitor, NRF2-IN-1, NRF2-IN1), 200mg, CAS: 846557-71-9.
SDP
Catalog No. 5000061037
Encompass_Preferred
ML385 (CAS 846557-71-9) is a selective inhibitor of the transcription factor nuclear factor erythroid 2-related factor 2 (NRF2) displaying an IC50 of 1 9 M NRF2 regulates cellular antioxidant responses detoxification pathways and multidrug transporters and its activation is frequently implicated in therapeutic resistance of non-small cell lung cancer (NSCLC) In A549 cells ML385 treatment reduces expression of NRF2-dependent genes dose- and time-dependently Additionally in NSCLC mouse models ML385 treatment diminished tumor growth and metastasis especially when combined with carboplatin underscoring its utility in cancer research and potential therapeutic exploration
Catalog No. 50-000-61037 Supplier Apexbio Technology LLC Supplier No. B8300200
ML385 (CAS 846557-71-9) is a selective inhibitor of the transcription factor nuclear factor erythroid 2-related factor 2 (NRF2) displaying an IC50 of 1 9 M NRF2 regulates cellular antioxidant responses detoxification pathways and multidrug transporters and its activation is frequently implicated in therapeutic resistance of non-small cell lung cancer (NSCLC) In A549 cells ML385 treatment reduces expression of NRF2-dependent genes dose- and time-dependently Additionally in NSCLC mouse models ML385 treatment diminished tumor growth and metastasis especially when combined with carboplatin underscoring its utility in cancer research and potential therapeutic exploration