Learn More
Selleck Chemical LLC ML385 5mg 846557-71-9
SDP
Supplier:  Selleck Chemical LLC S87905MG
Encompass_Preferred
ML385 is a novel and specific NRF2 inhibitor with an IC50 of 1.9 ?M. It inhibits the downstream target gene expression of NRF2. NRF2 regulates the activity of several ferroptosis and lipid peroxidation-related proteins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8057