Learn More
Medchemexpress LLC N-[3-oxo-3-[[(1R)-1,2,3,4-tetrahydro-1-naphthalenyl]amino]propyl]-4-(trifluoromethoxy)-benzamide | 2029049-79-2 | MFCD30489750 | 99.7% | 406.40 | C21H21F3N2O3 | 5 MG
SDP
Catalog No. 501967768
Encompass_Preferred
ML390 is a small-molecule inhibitor of dihydroorotate dehydrogenase (DHODH) that induces myeloid differentiation in murine and human acute myeloid leukemia (AML) models. It is supplied as a solid research-grade compound for biochemical and cellular studies.
  • Potent DHODH inhibitor that induces myeloid differentiation in AML models.
  • High purity (99.72%) suitable for biochemical and cellular assays.
  • Molecular weight 406.40 and formula C21H21F3N2O3 for accurate dosing and calculations.
  • White to off-white solid appearance, easy to handle and weigh.
  • Stable as a powder at -20°C for up to 3 years and at 4°C for up to 2 years.
  • Available in small research pack sizes including 5 mg for screening and pilot studies.
Catalog No. 50-196-7768 Supplier Medchemexpress LLC Supplier No. HY1006885MG
ML390 is a small-molecule inhibitor of dihydroorotate dehydrogenase (DHODH) that induces myeloid differentiation in murine and human acute myeloid leukemia (AML) models. It is supplied as a solid research-grade compound for biochemical and cellular studies.
  • Potent DHODH inhibitor that induces myeloid differentiation in AML models.
  • High purity (99.72%) suitable for biochemical and cellular assays.
  • Molecular weight 406.40 and formula C21H21F3N2O3 for accurate dosing and calculations.
  • White to off-white solid appearance, easy to handle and weigh.
  • Stable as a powder at -20°C for up to 3 years and at 4°C for up to 2 years.
  • Available in small research pack sizes including 5 mg for screening and pilot studies.