Learn More
TARGETMOL CHEMICALS INC ML400 5MG
SDP
Catalog No. 502580739
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML400 (CID73050863) is an allosteric inhibitor of LMPTP with an EC50 of 1uM. ML400 displays good cell-based activity and rodent pharmacokinetics. purity: 99%
Catalog No. 50-258-0739 Supplier TARGETMOL CHEMICALS INC Supplier No. T258245MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML400 (CID73050863) is an allosteric inhibitor of LMPTP with an EC50 of 1uM. ML400 displays good cell-based activity and rodent pharmacokinetics. purity: 99%