Learn More
Medchemexpress LLC HY-116814 10mg Medchemexpress, ML401 CAS:1597489-14-9 Purity:>98%
SDP
Catalog No. 501969441
Encompass_Preferred
Medchemexpress, HY-116814 10mg ML401 CAS:1597489-14-9 ML401, a potent chemical probe, selectively antagonizes EBI2 (also known as GPR183) with an IC50 of 1.03 nM. ML401 displays activity in a chemotaxis assay (IC50=6.24 nM). ML401 shows good stability and no toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9441 Supplier Medchemexpress LLC Supplier No. HY11681410MG
Medchemexpress, HY-116814 10mg ML401 CAS:1597489-14-9 ML401, a potent chemical probe, selectively antagonizes EBI2 (also known as GPR183) with an IC50 of 1.03 nM. ML401 displays activity in a chemotaxis assay (IC50=6.24 nM). ML401 shows good stability and no toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.