Learn More
Medchemexpress LLC HY-104027 5mg Medchemexpress, ML402 CAS:298684-44-3 Purity:>98%
SDP
Catalog No. 501968192
Encompass_Preferred
Medchemexpress, HY-104027 5mg ML402 CAS:298684-44-3 ML402 is a selective TREK-1 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8192 Supplier Medchemexpress LLC Supplier No. HY1040275MG
Medchemexpress, HY-104027 5mg ML402 CAS:298684-44-3 ML402 is a selective TREK-1 activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.