Learn More
Medchemexpress LLC HY-124416 5mg Medchemexpress, ML604086 CAS:850330-18-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1244165MG
Encompass_Preferred
Medchemexpress, HY-124416 5mg ML604086 CAS:850330-18-6 ML604086 is a selective CCR8 inhibitor, inhibiting CCL1 binding to CCR8 on circulating T-cells. ML604086 inhibits CCL1 mediated chemotaxis and increases in intracellular Ca2+ concentrations[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9991