Learn More
Medchemexpress LLC ML604086 | 850330-18-6 | 99.8% | 508.63 | C27H32N4O4S | 100MG
SDP
Catalog No. 5000237044
Encompass_Preferred
ML604086 is a selective CCR8 inhibitor that blocks CCL1 binding to CCR8 on circulating T cells, inhibiting CCL1-mediated chemotaxis and modulating intracellular Ca2+ increases. The compound is provided as a white to off-white solid with high reported purity and is intended for research use only.
  • Selective CCR8 inhibition for targeted immune signaling studies.
  • Blocks CCL1 binding and reduces chemotaxis in T-cell assays.
  • Modulates intracellular Ca2+ responses in functional assays.
  • High reported purity (99.78%), suitable for biochemical experiments.
  • Solid form, white to off-white, convenient for storage and formulation.
  • Available in multiple pack sizes including 100 mg for experimental flexibility.
Catalog No. 50-002-37044 Supplier Medchemexpress LLC Supplier No. HY124416100MG
ML604086 is a selective CCR8 inhibitor that blocks CCL1 binding to CCR8 on circulating T cells, inhibiting CCL1-mediated chemotaxis and modulating intracellular Ca2+ increases. The compound is provided as a white to off-white solid with high reported purity and is intended for research use only.
  • Selective CCR8 inhibition for targeted immune signaling studies.
  • Blocks CCL1 binding and reduces chemotaxis in T-cell assays.
  • Modulates intracellular Ca2+ responses in functional assays.
  • High reported purity (99.78%), suitable for biochemical experiments.
  • Solid form, white to off-white, convenient for storage and formulation.
  • Available in multiple pack sizes including 100 mg for experimental flexibility.