Learn More
Abcam ML7, myosin light chain kinase inhibitor, 50MG
Catalog No. 502707198
Encompass_Preferred
MW 452.7 Da, Purity >99%. Potent and selective myosin light chain kinase inhibitor (MLCK) (Ki = 300 nM). Also inhibits PKA and PKC (Ki values are 21 and 42 μM, respectively). Cell-permeable. Inhibits tumor cell growth.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7198 Supplier Abcam Supplier No. AB12084850MG
MW 452.7 Da, Purity >99%. Potent and selective myosin light chain kinase inhibitor (MLCK) (Ki = 300 nM). Also inhibits PKA and PKC (Ki values are 21 and 42 μM, respectively). Cell-permeable. Inhibits tumor cell growth.
The product is subject to the following: Abcam Restricted Use Statement